Assay Details
| Gene Symbol: fn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131520.2 | NP_571595.2 | 42 - 43 | 7155 | 65 | |
| XM_009304503.3 | XP_009302778.1 | 42 - 43 | 7189 | 65 | ||
| GenBank mRNA | AF081128.1 | - | 42 - 43 | 7101 | 65 | |
| BC162574.1 | - | 42 - 43 | 7094 | 65 | ||
More Information
Set Membership: |
|

fn1a
Genomic Map