Assay Details
| Gene Symbol: dvl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131757.2 | NP_571832.2 | 12 - 13 | 1716 | 86 | |
| GenBank mRNA | AB041734.1 | - | 12 - 13 | 1747 | 86 | |
| BC044353.1 | - | 12 - 13 | 1698 | 86 | ||
| BC098888.1 | - | 12 - 13 | 1689 | 86 | ||
| BC165644.1 | - | 12 - 13 | 1382 | 86 | ||
More Information
Set Membership: |
|

dvl3a
Genomic Map