Assay Details
| Gene Symbol: dvl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131757.2 | NP_571832.2 | 14 - 15 | 2074 | 87 | |
| GenBank mRNA | AB041734.1 | - | 14 - 15 | 87 | ||
| BC044353.1 | - | 14 - 15 | 87 | |||
| BC098888.1 | - | 14 - 15 | 87 | |||
| BC165644.1 | - | 14 - 15 | 87 | |||
More Information
Set Membership: |
|

dvl3a
Genomic Map