Assay Details
| Gene Symbol: cry5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131788.1 | NP_571863.1 | 3 - 4 | 525 | 103 | |
| XM_005155405.3 | XP_005155462.1 | 4 - 5 | 662 | 103 | ||
| GenBank mRNA | AB042254.1 | - | 3 - 4 | 525 | 103 | |
| BC044204.1 | - | 4 - 5 | 604 | 103 | ||
More Information
Set Membership: |
|

cry5
Genomic Map