Assay Details
| Gene Symbol: cry5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131788.1 | NP_571863.1 | 9 - 10 | 1499 | 83 | |
| XM_005155405.3 | XP_005155462.1 | 10 - 11 | 1636 | 83 | ||
| GenBank mRNA | AB042254.1 | - | 9 - 10 | 1499 | 83 | |
More Information
Set Membership: |
|

cry5
Genomic Map