Assay Details
| Gene Symbol: prickle2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183343.1 | NP_899186.1 | 6 - 7 | 854 | 95 | |
| XM_005155827.3 | XP_005155884.1 | 6 - 7 | 1170 | 95 | ||
| GenBank mRNA | AY278987.1 | - | 6 - 7 | 854 | 95 | |
| BC163236.1 | - | 6 - 7 | 912 | 95 | ||
| BC163238.1 | - | 6 - 7 | 912 | 95 | ||
More Information
Set Membership: |
|

prickle2b
Genomic Map