Assay Details
| Gene Symbol: cbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194373.1 | NP_919354.1 | 4 - 5 | 279 | 65 | |
| GenBank mRNA | AB074154.1 | - | 4 - 5 | 279 | 65 | |
| BC052140.1 | - | 4 - 5 | 300 | 65 | ||
| BC085553.1 | - | 4 - 5 | 320 | 65 | ||
| BC164860.1 | - | 4 - 5 | 291 | 65 | ||
More Information
Set Membership: |
|

cbx2
Genomic Map