Assay Details
| Gene Symbol: ndnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282089.1 | NP_001269018.1 | 8 - 9 | 799 | 97 | |
| NM_001282090.1 | NP_001269019.1 | 9 - 10 | 678 | 97 | ||
| GenBank mRNA | AY228339.1 | - | 9 - 10 | 657 | 97 | |
| AY288919.1 | - | 3 - 4 | 1226 | 98 | ||
| BC086747.1 | - | 9 - 10 | 686 | 97 | ||
| BC165513.1 | - | 8 - 9 | 619 | 97 | ||
| EH545346.1 | - | 9 - 10 | 703 | 97 | ||
| EH552618.1 | - | 8 - 9 | 833 | 97 | ||
| EH568837.1 | - | 2 - 3 | 415 | 97 | ||
More Information
Set Membership: |
|

ndnl2
Genomic Map