Assay Details
| Gene Symbol: fen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328507.1 | NP_001315436.1 | 2 - 3 | 215 | 107 | |
| XM_017353804.1 | XP_017209293.1 | 2 - 3 | 233 | 107 | ||
| GenBank mRNA | BC049413.1 | - | 2 - 3 | 154 | 107 | |
| CT612850.2 | - | 2 - 3 | 194 | 107 | ||
| EH439441.1 | - | 2 - 3 | 223 | 107 | ||
More Information
Set Membership: |
|

fen1
Genomic Map