Assay Details
| Gene Symbol: fen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328507.1 | NP_001315436.1 | 4 - 5 | 419 | 57 | |
| XM_017353804.1 | XP_017209293.1 | 4 - 5 | 437 | 57 | ||
| GenBank mRNA | AY391423.1 | - | 4 - 5 | 348 | 57 | |
| AY648844.1 | - | 4 - 5 | 361 | 57 | ||
| BC049413.1 | - | 4 - 5 | 358 | 57 | ||
| BC071488.1 | - | 4 - 5 | 380 | 57 | ||
| CT612850.2 | - | 4 - 5 | 398 | 57 | ||
| EH439441.1 | - | 4 - 5 | 427 | 57 | ||
More Information
Set Membership: |
|

fen1
Genomic Map