Assay Details
| Gene Symbol: rbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199528.3 | NP_955822.3 | 3 - 4 | 695 | 102 | |
| GenBank mRNA | AY309074.1 | - | 3 - 4 | 462 | 102 | |
| BC060936.1 | - | 3 - 4 | 631 | 102 | ||
| BC066514.1 | - | 3 - 4 | 635 | 102 | ||
| EH449687.1 | - | 3 - 4 | 664 | 102 | ||
| EH457906.1 | - | 1 - 2 | 103 | |||
| EH597149.1 | - | 1 - 2 | 600 | 102 | ||
| EH602992.1 | - | 1 - 2 | 600 | 102 | ||
More Information
Set Membership: |
|

rbp5
Genomic Map