Assay Details
| Gene Symbol: ssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199547.1 | NP_955841.1 | 9 - 10 | 895 | 63 | |
| XM_005172073.4 | XP_005172130.1 | 9 - 10 | 2982 | 63 | ||
| XM_009302102.3 | XP_009300377.1 | 10 - 11 | 3098 | 63 | ||
| GenBank mRNA | BC045392.1 | - | 9 - 10 | 895 | 63 | |
| BC165298.1 | - | 8 - 9 | 840 | 63 | ||
More Information
Set Membership: |
|

ssb
Genomic Map