Assay Details
| Gene Symbol: calm3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199570.1 | NP_955864.1 | 4 - 5 | 363 | 74 | |
| GenBank mRNA | BC044434.1 | - | 4 - 5 | 363 | 74 | |
| BC066752.1 | - | 4 - 5 | 395 | 74 | ||
More Information
Set Membership: |
|

calm3b
Genomic Map