Assay Details
| Gene Symbol: rhoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199818.1 | NP_956112.1 | 4 - 5 | 797 | 65 | |
| GenBank mRNA | BC045850.1 | - | 4 - 5 | 797 | 65 | |
| BC075783.1 | - | 4 - 5 | 798 | 65 | ||
| BC152679.1 | - | 4 - 5 | 797 | 65 | ||
| BC164028.1 | - | 4 - 5 | 501 | 65 | ||
More Information
Set Membership: |
|

rhoq
Genomic Map