Assay Details
| Gene Symbol: tdo2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199856.1 | NP_956150.1 | 7 - 8 | 792 | 115 | |
| XM_021479554.1 | XP_021335229.1 | 7 - 8 | 796 | 115 | ||
| GenBank mRNA | BC055123.1 | - | 7 - 8 | 792 | 115 | |
More Information
Set Membership: |
|

tdo2b
Genomic Map