Assay Details
| Gene Symbol: hspa4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199857.1 | NP_956151.1 | 14 - 15 | 1921 | 65 | |
| GenBank mRNA | BC048063.1 | - | 14 - 15 | 1921 | 65 | |
| BC065970.1 | - | 14 - 15 | 65 | |||
More Information
Set Membership: |
|

hspa4b
Genomic Map