Assay Details
| Gene Symbol: hspa4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199857.1 | NP_956151.1 | 8 - 9 | 1099 | 118 | |
| GenBank mRNA | BC048063.1 | - | 8 - 9 | 1099 | 118 | |
| BC065970.1 | - | 8 - 9 | 1128 | 118 | ||
More Information
Set Membership: |
|

hspa4b
Genomic Map