Assay Details
| Gene Symbol: abi3bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199859.1 | NP_956153.1 | 12 - 13 | 1435 | 87 | |
| GenBank mRNA | BC045333.1 | - | 12 - 13 | 1435 | 87 | |
| BC165666.1 | - | 12 - 13 | 1352 | 87 | ||
More Information
Set Membership: |
|

abi3bpb
Genomic Map