Assay Details
| Gene Symbol: abi3bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199859.1 | NP_956153.1 | 14 - 15 | 1649 | 92 | |
| XM_005161776.4 | XP_005161833.1 | 15 - 16 | 1727 | 92 | ||
| XM_005161777.4 | XP_005161834.1 | 14 - 15 | 1526 | 92 | ||
| XM_009296432.3 | XP_009294707.1 | 13 - 14 | 1493 | 92 | ||
| XM_009296433.3 | XP_009294708.1 | 13 - 14 | 1451 | 92 | ||
| XM_009296434.3 | XP_009294709.1 | 12 - 13 | 1379 | 92 | ||
| XM_009296436.3 | XP_009294711.1 | 11 - 12 | 1217 | 92 | ||
| XM_009296437.3 | XP_009294712.1 | 11 - 12 | 1211 | 92 | ||
| XM_009296438.3 | XP_009294713.1 | 10 - 11 | 1145 | 92 | ||
| XM_021469291.1 | XP_021324966.1 | 14 - 15 | 1652 | 92 | ||
| XM_021469292.1 | XP_021324967.1 | 12 - 13 | 1283 | 92 | ||
| GenBank mRNA | BC045333.1 | - | 14 - 15 | 1649 | 92 | |
| BC165666.1 | - | 14 - 15 | 1566 | 92 | ||
More Information
Set Membership: |
|

abi3bpb
Genomic Map