Assay Details
| Gene Symbol: abi3bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199859.1 | NP_956153.1 | 19 - 20 | 2237 | 72 | |
| XM_005161776.4 | XP_005161833.1 | 20 - 21 | 2315 | 72 | ||
| XM_005161777.4 | XP_005161834.1 | 19 - 20 | 2114 | 72 | ||
| XM_009296432.3 | XP_009294707.1 | 18 - 19 | 2081 | 72 | ||
| XM_009296433.3 | XP_009294708.1 | 18 - 19 | 2039 | 72 | ||
| XM_009296434.3 | XP_009294709.1 | 17 - 18 | 1967 | 72 | ||
| XM_009296436.3 | XP_009294711.1 | 16 - 17 | 1805 | 72 | ||
| XM_009296437.3 | XP_009294712.1 | 16 - 17 | 1799 | 72 | ||
| XM_009296438.3 | XP_009294713.1 | 15 - 16 | 1733 | 72 | ||
| XM_021469291.1 | XP_021324966.1 | 19 - 20 | 2240 | 72 | ||
| XM_021469292.1 | XP_021324967.1 | 17 - 18 | 1871 | 72 | ||
| GenBank mRNA | BC045333.1 | - | 19 - 20 | 2237 | 72 | |
| BC165666.1 | - | 19 - 20 | 2154 | 72 | ||
More Information
Set Membership: |
|

abi3bpb
Genomic Map