Assay Details
| Gene Symbol: abi3bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199859.1 | NP_956153.1 | 7 - 8 | 858 | 67 | |
| XM_005161776.4 | XP_005161833.1 | 7 - 8 | 870 | 67 | ||
| XM_005161777.4 | XP_005161834.1 | 7 - 8 | 870 | 67 | ||
| XM_009296432.3 | XP_009294707.1 | 7 - 8 | 870 | 67 | ||
| XM_009296433.3 | XP_009294708.1 | 7 - 8 | 870 | 67 | ||
| XM_009296436.3 | XP_009294711.1 | 7 - 8 | 870 | 67 | ||
| XM_021469291.1 | XP_021324966.1 | 7 - 8 | 870 | 67 | ||
| XM_021469292.1 | XP_021324967.1 | 7 - 8 | 870 | 67 | ||
| GenBank mRNA | BC045333.1 | - | 7 - 8 | 858 | 67 | |
| BC165666.1 | - | 7 - 8 | 775 | 67 | ||
More Information
Set Membership: |
|

abi3bpb
Genomic Map