Assay Details
| Gene Symbol: abi3bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199859.1 | NP_956153.1 | 9 - 10 | 1005 | 77 | |
| XM_005161776.4 | XP_005161833.1 | 9 - 10 | 1017 | 77 | ||
| XM_009296432.3 | XP_009294707.1 | 9 - 10 | 1017 | 77 | ||
| GenBank mRNA | BC045333.1 | - | 9 - 10 | 1005 | 77 | |
| BC165666.1 | - | 9 - 10 | 922 | 77 | ||
More Information
Set Membership: |
|

abi3bpb
Genomic Map