Assay Details
| Gene Symbol: gas6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199978.1 | NP_956272.1 | 12 - 13 | 1515 | 70 | |
| GenBank mRNA | BC053117.1 | - | 12 - 13 | 1515 | 70 | |
More Information
Set Membership: |
|

gas6
Genomic Map