Assay Details
| Gene Symbol: kidins220b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199982.2 | NP_956276.2 | 4 - 5 | 486 | 95 | |
| XM_005160637.4 | XP_005160694.1 | 4 - 5 | 525 | 95 | ||
| XM_005160638.4 | XP_005160695.1 | 4 - 5 | 525 | 95 | ||
| XM_005160639.4 | XP_005160696.1 | 4 - 5 | 525 | 95 | ||
| XM_005160640.4 | XP_005160697.1 | 4 - 5 | 525 | 95 | ||
| GenBank mRNA | BC061450.1 | - | 4 - 5 | 486 | 95 | |
More Information
Set Membership: |
|

kidins220b
Genomic Map