Assay Details
| Gene Symbol: serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200184.1 | NP_956478.1 | 7 - 8 | 1152 | 60 | |
| GenBank mRNA | BC045498.1 | - | 7 - 8 | 1152 | 60 | |
| BC163961.1 | - | 6 - 7 | 1094 | 60 | ||
More Information
Set Membership: |
|

serpine2
Genomic Map