Assay Details
| Gene Symbol: eps8l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328079.1 | NP_001315008.1 | 4 - 5 | 291 | 81 | |
| XM_005167020.4 | XP_005167077.1 | 4 - 5 | 596 | 81 | ||
| GenBank mRNA | BC053196.1 | - | 3 - 4 | 195 | 81 | |
More Information
Set Membership: |
|

eps8l3b
Genomic Map