Assay Details
| Gene Symbol: eps8l3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328079.1 | NP_001315008.1 | 9 - 10 | 937 | 71 | |
| XM_005167020.4 | XP_005167077.1 | 9 - 10 | 1242 | 71 | ||
| GenBank mRNA | BC053196.1 | - | 8 - 9 | 841 | 71 | |
More Information
Set Membership: |
|

eps8l3b
Genomic Map