Assay Details
| Gene Symbol: slc25a39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200486.1 | NP_956780.1 | 11 - 12 | 1069 | 99 | |
| XM_005163929.4 | XP_005163986.1 | 11 - 12 | 1102 | 99 | ||
| XM_005163930.4 | XP_005163987.1 | 10 - 11 | 1049 | 99 | ||
| XM_005163931.4 | XP_005163988.1 | 10 - 11 | 1046 | 99 | ||
| XM_021469806.1 | XP_021325481.1 | 11 - 12 | 1093 | 99 | ||
| GenBank mRNA | BC055226.1 | - | 11 - 12 | 1069 | 99 | |
More Information
Set Membership: |
|

slc25a39
Genomic Map