Assay Details
| Gene Symbol: srsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200533.2 | NP_956827.1 | 4 - 5 | 539 | 77 | |
| GenBank mRNA | BC056275.1 | - | 4 - 5 | 523 | 77 | |
| BC066442.1 | - | 4 - 5 | 539 | 77 | ||
More Information
Set Membership: |
|

srsf10b
Genomic Map