Assay Details
| Gene Symbol: slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200681.1 | NP_956975.1 | 9 - 10 | 1048 | 62 | |
| GenBank mRNA | BC058316.1 | - | 9 - 10 | 1048 | 62 | |
| BC067621.1 | - | 9 - 10 | 1058 | 62 | ||
More Information
Set Membership: |
|

slc5a1
Genomic Map