Assay Details
| Gene Symbol: ndrg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200692.3 | NP_956986.2 | 11 - 12 | 929 | 115 | |
| GenBank mRNA | BC059429.1 | - | 11 - 12 | 874 | 115 | |
More Information
Set Membership: |
|

ndrg1b
Genomic Map