Assay Details
| Gene Symbol: msi2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201109.1 | NP_957403.1 | 13 - 14 | 1209 | 97 | |
| XM_005157680.4 | XP_005157737.1 | 13 - 14 | 1324 | 97 | ||
| XM_005157681.4 | XP_005157738.1 | 13 - 14 | 1325 | 97 | ||
| XM_005157682.4 | XP_005157739.1 | 13 - 14 | 1266 | 97 | ||
| XM_005157685.4 | XP_005157742.1 | 12 - 13 | 1200 | 97 | ||
| XM_005157686.4 | XP_005157743.1 | 12 - 13 | 1085 | 97 | ||
| XM_005157687.4 | XP_005157744.1 | 11 - 12 | 1018 | 97 | ||
| XM_009291633.3 | XP_009289908.1 | 12 - 13 | 1144 | 97 | ||
| GenBank mRNA | BC055251.1 | - | 13 - 14 | 1209 | 97 | |
More Information
Set Membership: |
|

msi2b
Genomic Map