Assay Details
| Gene Symbol: msi2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201109.1 | NP_957403.1 | 14 - 15 | 1286 | 98 | |
| GenBank mRNA | BC055251.1 | - | 14 - 15 | 1286 | 98 | |
More Information
Set Membership: |
|

msi2b
Genomic Map