Assay Details
| Gene Symbol: atp6v1aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201135.2 | NP_957429.1 | 11 - 12 | 1356 | 86 | |
| GenBank mRNA | BC055130.1 | - | 11 - 12 | 1357 | 86 | |
More Information
Set Membership: |
|

atp6v1aa
Genomic Map