Assay Details
| Gene Symbol: bub3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205709.1 | NP_991272.1 | 3 - 4 | 413 | 75 | |
| XM_005169505.4 | XP_005169562.1 | 4 - 5 | 561 | 75 | ||
| GenBank mRNA | AY391411.1 | - | 3 - 4 | 413 | 75 | |
| BC083205.1 | - | 4 - 5 | 477 | 75 | ||
More Information
Set Membership: |
|

bub3
Genomic Map