Assay Details
| Gene Symbol: pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205718.3 | NP_991281.3 | 5 - 6 | 809 | 66 | |
| GenBank mRNA | AJ579536.1 | - | 5 - 6 | 770 | 66 | |
| BC095289.1 | - | 5 - 6 | 765 | 66 | ||
| BC165557.1 | - | 5 - 6 | 784 | 66 | ||
| EH555457.1 | - | 5 - 6 | 853 | 66 | ||
| EH588556.1 | - | 3 - 4 | 390 | 66 | ||
More Information
Set Membership: |
|

pofut1
Genomic Map