Assay Details
| Gene Symbol: leo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207067.1 | NP_996950.1 | 7 - 8 | 1405 | 60 | |
| GenBank mRNA | BC066443.1 | - | 7 - 8 | 1405 | 60 | |
More Information
Set Membership: |
|

leo1
Genomic Map