Assay Details
| Gene Symbol: ilf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212599.1 | NP_997764.1 | 8 - 9 | 959 | 81 | |
| XM_005163739.4 | XP_005163796.1 | 8 - 9 | 1031 | 81 | ||
| XM_005163740.4 | XP_005163797.1 | 8 - 9 | 1031 | 81 | ||
| XM_005163741.3 | XP_005163798.1 | 8 - 9 | 1028 | 81 | ||
| XM_005163742.4 | XP_005163799.1 | 8 - 9 | 1028 | 81 | ||
| XM_009299225.3 | XP_009297500.1 | 7 - 8 | 939 | 81 | ||
| GenBank mRNA | BC047175.1 | - | 8 - 9 | 988 | 81 | |
| BC067172.1 | - | 8 - 9 | 959 | 81 | ||
More Information
Set Membership: |
|

ilf3b
Genomic Map