Assay Details
| Gene Symbol: eif2s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212711.3 | NP_997876.2 | 3 - 4 | 562 | 85 | |
| GenBank mRNA | AY648723.1 | - | 3 - 4 | 562 | 85 | |
| BC055248.1 | - | 3 - 4 | 296 | 85 | ||
More Information
Set Membership: |
|

eif2s3
Genomic Map