Assay Details
| Gene Symbol: irf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327817.1 | NP_001314746.1 | 3 - 4 | 514 | 61 | |
| GenBank mRNA | BC066751.1 | - | 3 - 4 | 399 | 61 | |
| EU274624.1 | - | 2 - 3 | 178 | 61 | ||
More Information
Set Membership: |
|

irf5
Genomic Map