Assay Details
| Gene Symbol: nrp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212965.1 | NP_998130.1 | 12 - 13 | 2650 | 90 | |
| XM_021469092.1 | XP_021324767.1 | 12 - 13 | 2670 | 90 | ||
| GenBank mRNA | AY351912.1 | - | 12 - 13 | 2386 | 90 | |
| AY428801.1 | - | 12 - 13 | 2650 | 90 | ||
| AY437405.1 | - | 12 - 13 | 2636 | 90 | ||
| BC162118.1 | - | 12 - 13 | 2532 | 90 | ||
More Information
Set Membership: |
|

nrp2a
Genomic Map