Assay Details
| Gene Symbol: nrp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212965.1 | NP_998130.1 | 4 - 5 | 1284 | 128 | |
| XM_021469092.1 | XP_021324767.1 | 4 - 5 | 1304 | 128 | ||
| GenBank mRNA | AY428801.1 | - | 4 - 5 | 1284 | 128 | |
| AY437399.1 | - | 4 - 5 | 668 | 128 | ||
| AY437405.1 | - | 4 - 5 | 1270 | 128 | ||
| BC162118.1 | - | 4 - 5 | 1166 | 128 | ||
More Information
Set Membership: |
|

nrp2a
Genomic Map