Assay Details
| Gene Symbol: spint1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213152.1 | NP_998317.1 | 6 - 7 | 1104 | 96 | |
| XM_021467136.1 | XP_021322811.1 | 7 - 8 | 1192 | 96 | ||
| XM_021467137.1 | XP_021322812.1 | 7 - 8 | 1193 | 96 | ||
| GenBank mRNA | AY648768.1 | - | 6 - 7 | 1128 | 96 | |
| BC053239.1 | - | 6 - 7 | 1104 | 96 | ||
| BC155204.1 | - | 6 - 7 | 1053 | 96 | ||
| BC165390.1 | - | 5 - 6 | 981 | 96 | ||
More Information
Set Membership: |
|

spint1a
Genomic Map