Assay Details
| Gene Symbol: spint1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213152.1 | NP_998317.1 | 9 - 10 | 1448 | 74 | |
| XM_021467136.1 | XP_021322811.1 | 10 - 11 | 1536 | 74 | ||
| XM_021467137.1 | XP_021322812.1 | 9 - 10 | 1486 | 74 | ||
| GenBank mRNA | AY648768.1 | - | 9 - 10 | 1472 | 74 | |
| BC053239.1 | - | 9 - 10 | 1448 | 74 | ||
| BC155204.1 | - | 9 - 10 | 1397 | 74 | ||
| BC165390.1 | - | 8 - 9 | 1325 | 74 | ||
More Information
Set Membership: |
|

spint1a
Genomic Map