Assay Details
| Gene Symbol: aldoab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213215.1 | NP_998380.1 | 2 - 3 | 146 | 84 | |
| GenBank mRNA | BC065847.1 | - | 2 - 3 | 146 | 84 | |
More Information
Set Membership: |
|

aldoab
Genomic Map