Assay Details
| Gene Symbol: rhoac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213350.1 | NP_998515.1 | 3 - 4 | 310 | 96 | |
| GenBank mRNA | BC054576.1 | - | 3 - 4 | 310 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rhoac| Gene Symbol: rhoac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213350.1 | NP_998515.1 | 3 - 4 | 310 | 96 | |
| GenBank mRNA | BC054576.1 | - | 3 - 4 | 310 | 96 | |
Set Membership: |
|