Assay Details
| Gene Symbol: spg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213395.1 | NP_998560.1 | 5 - 6 | 641 | 61 | |
| GenBank mRNA | AY394947.1 | - | 5 - 6 | 710 | 61 | |
| BC059461.1 | - | 5 - 6 | 641 | 61 | ||
More Information
Set Membership: |
|

spg21
Genomic Map