Assay Details
| Gene Symbol: slc25a55a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213408.1 | NP_998573.1 | 6 - 7 | 580 | 64 | |
| XM_005172105.4 | XP_005172162.1 | 6 - 7 | 589 | 64 | ||
| XM_005172109.3 | XP_005172166.1 | 7 - 8 | 657 | 64 | ||
| XM_009302668.2 | XP_009300943.1 | 7 - 8 | 640 | 64 | ||
| XM_017356461.1 | XP_017211950.1 | 6 - 7 | 615 | 64 | ||
| GenBank mRNA | BC051620.1 | - | 6 - 7 | 580 | 64 | |
| BC075788.1 | - | 6 - 7 | 583 | 64 | ||
More Information
Set Membership: |
|

slc25a55a
Genomic Map