Assay Details
| Gene Symbol: slc25a55a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213408.1 | NP_998573.1 | 8 - 9 | 924 | 66 | |
| XM_005172105.4 | XP_005172162.1 | 8 - 9 | 933 | 66 | ||
| XM_005172109.3 | XP_005172166.1 | 9 - 10 | 1001 | 66 | ||
| XM_009302668.2 | XP_009300943.1 | 9 - 10 | 984 | 66 | ||
| XM_017356461.1 | XP_017211950.1 | 8 - 9 | 959 | 66 | ||
| GenBank mRNA | BC051620.1 | - | 8 - 9 | 924 | 66 | |
| BC075788.1 | - | 8 - 9 | 927 | 66 | ||
More Information
Set Membership: |
|

slc25a55a
Genomic Map