Assay Details
| Gene Symbol: pitpnbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213443.1 | NP_998608.1 | 7 - 8 | 589 | 91 | |
| GenBank mRNA | BC046882.1 | - | 7 - 8 | 589 | 91 | |
| BC164619.1 | - | 7 - 8 | 488 | 91 | ||
More Information
Set Membership: |
|

pitpnbl
Genomic Map